Learn More
Enzo Life Sciences Dopamine transporter (extracellular loop) polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLSA5090100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to a portion of the extracellular loop 2 (EL2) region of human dopamine transporter. Species reactivity: Human, Mouse Monkey. Applications: IHC, WB. Formulation: Liquid. In 10mM HEPES, pH 7.5, containing 150mM NaCl, 10% BSA and 50% glycerol.
Catalog No. 50-201-1842