Learn More
Enzo Life Sciences SCREEN-WELL Dopaminergic ligand library (500 µl/well)
SDP
Catalog No. 502010191
Encompass_Preferred
The Dopaminergic Ligands Plate contains 80 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Dopaminergic Ligands plate is 1 plate of the larger Neurotransmitter Library (BML-2810) which contains over 650 ligands. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-0191 Supplier Enzo Life Sciences Supplier No. BML28120500
The Dopaminergic Ligands Plate contains 80 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Dopaminergic Ligands plate is 1 plate of the larger Neurotransmitter Library (BML-2810) which contains over 650 ligands. Long Term Storage: -80°C. Applications: HTS .