Learn More
Cayman Chemical Dopamine (hydrochloride), Purity: ≥98%, CAS: 62-31-7, 25 g
SDP
Catalog No. 502964198
Encompass_Preferred
An endogenous catecholamine neurotransmitter synthesized from L-tyrosine; agonist at D1-5 receptors; synthesized in the substantia nigra and ventral tegmental area; a precursor in norepinephrine and epinephrine biosynthesis; functions as a hormone in the adrenal medulla to regulate heart rate and blood pressure
Catalog No. 50-296-4198 Supplier Cayman Chemical Supplier No. 2199225 G
An endogenous catecholamine neurotransmitter synthesized from L-tyrosine; agonist at D1-5 receptors; synthesized in the substantia nigra and ventral tegmental area; a precursor in norepinephrine and epinephrine biosynthesis; functions as a hormone in the adrenal medulla to regulate heart rate and blood pressure