Learn More
Kyfora Bio DOPC 25 MG
Catalog No. 502559774
Encompass_Preferred
DOPC is a neutral helper phospholipid used in gene and drug delivery liposomal formulations. DOPC has been shown to promote more stable laminar structures. When used in combination with charged lipids, helper lipids have been shown to increase the uptake efficiency in a variety of cell types.Key application(s): LNP, liposomes
Catalog No. 50-255-9774 Supplier Kyfora Bio Supplier No. 2645025
DOPC is a neutral helper phospholipid used in gene and drug delivery liposomal formulations. DOPC has been shown to promote more stable laminar structures. When used in combination with charged lipids, helper lipids have been shown to increase the uptake efficiency in a variety of cell types.Key application(s): LNP, liposomes