Learn More
TARGETMOL CHEMICALS INC DOPE 250MG
SDP
Catalog No. 502581057
Encompass_Preferred
Also available in 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DOPE (DOPE) is a neutral helper lipid for cationic liposomes.It is used in combination with cationic phospholipids to increase efficiency during DNA transfection studies as a non-viral method of gene delivery. purity: 98%
Catalog No. 50-258-1057 Supplier TARGETMOL CHEMICALS INC Supplier No. T19080250MG
Also available in 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DOPE (DOPE) is a neutral helper lipid for cationic liposomes.It is used in combination with cationic phospholipids to increase efficiency during DNA transfection studies as a non-viral method of gene delivery. purity: 98%