Learn More
Medchemexpress LLC DOPE | 4004-05-1 | 99.96% | 744.03 | 25 MG
SDP
Catalog No. 5000438727
Encompass_Preferred
DOPE (Dioleoylphosphatidylethanolamine) is a neutral helper lipid for cationic liposomes, designed to improve the transfection efficiency of naked siRNA when combined with cationic phospholipids. This product is for research use only. In studies, DOPE, as a neutral lipid in lipid nanoparticles (LNP), has been shown to exert a knock-down effect on macrophages and DCs, and liposomes containing DOPE exhibit fusogenicity. LNP/siRNA significantly suppresses CD45 protein expression in macrophages and DCs, demonstrating its potential as an efficient siRNA delivery system for both murine and human macrophages and DCs.
  • Neutral helper lipid for cationic liposomes.
  • Improves transfection efficiency of naked siRNA.
  • Exerts knock-down effect on macrophages and DCs.
  • Shows fusogenicity.
  • Potential for efficient siRNA delivery system.
Catalog No. 50-004-38727 Supplier Medchemexpress LLC Supplier No. 25MGHY112005
DOPE (Dioleoylphosphatidylethanolamine) is a neutral helper lipid for cationic liposomes, designed to improve the transfection efficiency of naked siRNA when combined with cationic phospholipids. This product is for research use only. In studies, DOPE, as a neutral lipid in lipid nanoparticles (LNP), has been shown to exert a knock-down effect on macrophages and DCs, and liposomes containing DOPE exhibit fusogenicity. LNP/siRNA significantly suppresses CD45 protein expression in macrophages and DCs, demonstrating its potential as an efficient siRNA delivery system for both murine and human macrophages and DCs.
  • Neutral helper lipid for cationic liposomes.
  • Improves transfection efficiency of naked siRNA.
  • Exerts knock-down effect on macrophages and DCs.
  • Shows fusogenicity.
  • Potential for efficient siRNA delivery system.