Learn More
Medchemexpress LLC DOPE | 4004-05-1 | 99.96% | 744.03 | 50 MG
SDP
Catalog No. 5000439121
Encompass_Preferred
DOPE (Dioleoylphosphatidylethanolamine) is a neutral helper lipid that combines with cationic phospholipids to improve the transfection efficiency of naked siRNA, and it is also an endogenous metabolite. In studies, DOPE-containing liposomes (LNP) exhibited a knock-down (KD) effect on macrophages and dendritic cells and showed fusogenicity. LNP/siRNA significantly suppressed CD45 protein expression, demonstrating potential as an efficient siRNA delivery system for both murine and human macrophages and dendritic cells.
  • Neutral helper lipid for cationic liposomes
  • Improves transfection efficiency of naked siRNA
  • Exerts knock-down (KD) effect on macrophages and dendritic cells
  • Liposomes containing DOPE show fusogenicity
  • Potential for efficient siRNA delivery system
  • Endogenous metabolite
Catalog No. 50-004-39121 Supplier Medchemexpress LLC Supplier No. HY11200550MG
DOPE (Dioleoylphosphatidylethanolamine) is a neutral helper lipid that combines with cationic phospholipids to improve the transfection efficiency of naked siRNA, and it is also an endogenous metabolite. In studies, DOPE-containing liposomes (LNP) exhibited a knock-down (KD) effect on macrophages and dendritic cells and showed fusogenicity. LNP/siRNA significantly suppressed CD45 protein expression, demonstrating potential as an efficient siRNA delivery system for both murine and human macrophages and dendritic cells.
  • Neutral helper lipid for cationic liposomes
  • Improves transfection efficiency of naked siRNA
  • Exerts knock-down (KD) effect on macrophages and dendritic cells
  • Liposomes containing DOPE show fusogenicity
  • Potential for efficient siRNA delivery system
  • Endogenous metabolite