Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DOPE-GA | 228706-30-7 | C46H84NO11P | 25 MG
SDP

Catalog No. 5000375962
Encompass_Preferred

DOPE-GA is a lipid used in the preparation of liposomes for research on drug delivery and other applications. It is a key raw material for preparing liposome ELL-12, a non-myelosuppressive antiproliferative agent with anti-tumor activity. This product is intended for research use only.

  • Purity of 98.0% or greater
  • Oil appearance, colorless to light yellow
  • Soluble in DMSO (100 mg/mL)
  • Soluble in various in vivo protocols for research
  • Pure form storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-75962 Supplier Medchemexpress LLC Supplier No. 25MGHY150240
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DOPE-GA is a lipid used in the preparation of liposomes for research on drug delivery and other applications. It is a key raw material for preparing liposome ELL-12, a non-myelosuppressive antiproliferative agent with anti-tumor activity. This product is intended for research use only.

  • Purity of 98.0% or greater
  • Oil appearance, colorless to light yellow
  • Soluble in DMSO (100 mg/mL)
  • Soluble in various in vivo protocols for research
  • Pure form storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.