Learn More
Medchemexpress LLC Dope-mPEG | 95.0% | 2000 Da (average) | 50 MG
SDP
Catalog No. 5000280668
Encompass_Preferred
DOPE-mPEG (average PEG 2000 Da) is a PEGylated phosphatidylethanolamine lipid used to prepare sterically stabilized liposomes and giant unilamellar vesicles. The methoxy-PEG conjugation provides surface PEGylation that reduces pH sensitivity and improves colloidal stability. Supplied as a powder with reported high purity, it is intended for research use in liposomal formulation and membrane studies.
  • Provides steric stabilization to liposomal formulations
  • Reduces pH sensitivity of lipid bilayers
  • Suitable for preparation of targeted liposomal drug delivery and GUVs
  • Powder form for convenient storage and handling
  • Reported purity ≥95.0%
Catalog No. 50-002-80668 Supplier Medchemexpress LLC Supplier No. HYW44098850MG
DOPE-mPEG (average PEG 2000 Da) is a PEGylated phosphatidylethanolamine lipid used to prepare sterically stabilized liposomes and giant unilamellar vesicles. The methoxy-PEG conjugation provides surface PEGylation that reduces pH sensitivity and improves colloidal stability. Supplied as a powder with reported high purity, it is intended for research use in liposomal formulation and membrane studies.
  • Provides steric stabilization to liposomal formulations
  • Reduces pH sensitivity of lipid bilayers
  • Suitable for preparation of targeted liposomal drug delivery and GUVs
  • Powder form for convenient storage and handling
  • Reported purity ≥95.0%