Learn More
Medchemexpress LLC DOPE-NHS | 1010188-79-0 | C50H87N2O13P | 25 MG
SDP
Catalog No. 5000374324
Encompass_Preferred
DOPE-NHS is a linker used in research for conjugating peptides to exosomes and potentially other membrane-based nanoparticles, including applications in drug delivery. It has been utilized to conjugate rabies viral glycoprotein to mesenchymal stem cell-derived exosomes to address memory deficits by modulating inflammatory responses in an Alzheimer's disease mouse model. Additionally, DOPE-NHS has been employed to link cardiac homing peptides to exosomes for targeted delivery to myocardial infarction.
  • Allows peptides to be conjugated to exosomes
  • Can be used for membrane-based nanoparticles
  • Applicable in drug delivery research
  • Facilitates conjugation of MSC-derived exosomes with rabies viral glycoprotein
  • Enables linking of cardiac homing peptides to exosomes for targeted delivery
Catalog No. 50-003-74324 Supplier Medchemexpress LLC Supplier No. 25MGHY150241
DOPE-NHS is a linker used in research for conjugating peptides to exosomes and potentially other membrane-based nanoparticles, including applications in drug delivery. It has been utilized to conjugate rabies viral glycoprotein to mesenchymal stem cell-derived exosomes to address memory deficits by modulating inflammatory responses in an Alzheimer's disease mouse model. Additionally, DOPE-NHS has been employed to link cardiac homing peptides to exosomes for targeted delivery to myocardial infarction.
  • Allows peptides to be conjugated to exosomes
  • Can be used for membrane-based nanoparticles
  • Applicable in drug delivery research
  • Facilitates conjugation of MSC-derived exosomes with rabies viral glycoprotein
  • Enables linking of cardiac homing peptides to exosomes for targeted delivery