Learn More
Medchemexpress LLC DOPE-NHS | 1010188-79-0 | C50H87N2O13P | 250 MG
SDP
Catalog No. 5000374364
Encompass_Preferred
DOPE-NHS is a linker that facilitates the conjugation of peptides to exosomes and other membrane-based nanoparticles, making it suitable for drug delivery applications.
  • Can be used to conjugate peptides to exosomes.
  • Enables conjugation to other membrane-based nanoparticles.
  • Suitable for drug delivery applications.
  • Facilitates conjugation of MSC-derived exosomes with rabies viral glycoprotein.
  • Allows conjugation of exosomes with cardiac homing peptide.
Catalog No. 50-003-74364 Supplier Medchemexpress LLC Supplier No. 250MGHY150241
DOPE-NHS is a linker that facilitates the conjugation of peptides to exosomes and other membrane-based nanoparticles, making it suitable for drug delivery applications.
  • Can be used to conjugate peptides to exosomes.
  • Enables conjugation to other membrane-based nanoparticles.
  • Suitable for drug delivery applications.
  • Facilitates conjugation of MSC-derived exosomes with rabies viral glycoprotein.
  • Allows conjugation of exosomes with cardiac homing peptide.