Learn More
Medchemexpress LLC DOPE-NHS | 1010188-79-0 | C50H87N2O13P | 5 MG
SDP
Catalog No. 5000375325
Encompass_Preferred
DOPE-NHS is a linker used for conjugating peptides to exosomes and other membrane-based nanoparticles. It is also suitable for drug delivery applications.
  • Allows peptides to be conjugated to exosomes and membrane-based nanoparticles
  • Can be used for drug delivery
  • Facilitates conjugation of MSC derived exosomes with rabies viral glycoprotein
  • Enables conjugation of exosomes with cardiac homing peptide
Catalog No. 50-003-75325 Supplier Medchemexpress LLC Supplier No. HY1502415MG
DOPE-NHS is a linker used for conjugating peptides to exosomes and other membrane-based nanoparticles. It is also suitable for drug delivery applications.
  • Allows peptides to be conjugated to exosomes and membrane-based nanoparticles
  • Can be used for drug delivery
  • Facilitates conjugation of MSC derived exosomes with rabies viral glycoprotein
  • Enables conjugation of exosomes with cardiac homing peptide