Learn More
Medchemexpress LLC DOPE-NHS | 1010188-79-0 | C50H87N2O13P | 50 MG
SDP
Catalog No. 5000375578
Encompass_Preferred
DOPE-NHS is a linker used for conjugating peptides to exosomes and other membrane-based nanoparticles, and for drug delivery applications. It has been utilized to conjugate mesenchymal stem cell (MSC)-derived exosomes with rabies viral glycoprotein and exosomes with cardiac homing peptide.
  • Can be used for conjugating peptides to exosomes
  • Can be used for conjugating peptides to other membrane-based nanoparticles
  • Utilized for drug delivery purposes
  • Used to conjugate mesenchymal stem cell (MSC)-derived exosomes with rabies viral glycoprotein
  • Used to conjugate exosomes with cardiac homing peptide
Catalog No. 50-003-75578 Supplier Medchemexpress LLC Supplier No. HY15024150MG
DOPE-NHS is a linker used for conjugating peptides to exosomes and other membrane-based nanoparticles, and for drug delivery applications. It has been utilized to conjugate mesenchymal stem cell (MSC)-derived exosomes with rabies viral glycoprotein and exosomes with cardiac homing peptide.
  • Can be used for conjugating peptides to exosomes
  • Can be used for conjugating peptides to other membrane-based nanoparticles
  • Utilized for drug delivery purposes
  • Used to conjugate mesenchymal stem cell (MSC)-derived exosomes with rabies viral glycoprotein
  • Used to conjugate exosomes with cardiac homing peptide