Learn More
Medchemexpress LLC Dope-PEG-azide | 99.6% | 1000 (average) | 10 MG
SDP
Catalog No. 5000284779
Encompass_Preferred
DOPE-PEG-Azide (MW 1000) is a PEGylated dioleoyl phosphatidylethanolamine bearing a terminal azide group used as a functional lipid in liposome formulation and drug delivery research. It provides an azide handle for bioorthogonal conjugation while contributing amphiphilic character to lipid assemblies.
  • Provides azide functional group for click chemistry conjugation.
  • Average PEG molecular weight 1000 for spacer functionality.
  • High reported purity (≈99.6%).
  • Amphiphilic lipid suitable for liposome and nanoparticle incorporation.
  • Oil form, colorless to light yellow, soluble in DMSO (200 mg/mL).
  • Store at -20°C in pure form per manufacturer recommendations.
Catalog No. 50-002-84779 Supplier Medchemexpress LLC Supplier No. HYW59144810MG
DOPE-PEG-Azide (MW 1000) is a PEGylated dioleoyl phosphatidylethanolamine bearing a terminal azide group used as a functional lipid in liposome formulation and drug delivery research. It provides an azide handle for bioorthogonal conjugation while contributing amphiphilic character to lipid assemblies.
  • Provides azide functional group for click chemistry conjugation.
  • Average PEG molecular weight 1000 for spacer functionality.
  • High reported purity (≈99.6%).
  • Amphiphilic lipid suitable for liposome and nanoparticle incorporation.
  • Oil form, colorless to light yellow, soluble in DMSO (200 mg/mL).
  • Store at -20°C in pure form per manufacturer recommendations.