Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dope-peg-azide | 99.6% | 1000 (average) | 250 MG
SDP

Catalog No. 5000284260
Encompass_Preferred

DOPE-PEG-Azide (PEG MW 1000) is a phospholipid-PEG conjugate used as a biocompatible amphiphilic component in drug delivery and formulation research. It is supplied as a colorless to light yellow oil, characterized by high purity and stability under recommended storage conditions.

  • Purity 99.62%.
  • Average PEG molecular weight 1,000.
  • Form: oil; colorless to light yellow.
  • Useful for liposome and micelle formulation and azide-alkyne click bioconjugation.
  • Available in laboratory packaging including 250 mg quantities.
  • Storage recommendations: pure form -20°C; in solvent -80°C for 6 months or -20°C for 1 month.
  • Intended for research use only.

Catalog No. 50-002-84260 Supplier Medchemexpress LLC Supplier No. HYW591448250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DOPE-PEG-Azide (PEG MW 1000) is a phospholipid-PEG conjugate used as a biocompatible amphiphilic component in drug delivery and formulation research. It is supplied as a colorless to light yellow oil, characterized by high purity and stability under recommended storage conditions.

  • Purity 99.62%.
  • Average PEG molecular weight 1,000.
  • Form: oil; colorless to light yellow.
  • Useful for liposome and micelle formulation and azide-alkyne click bioconjugation.
  • Available in laboratory packaging including 250 mg quantities.
  • Storage recommendations: pure form -20°C; in solvent -80°C for 6 months or -20°C for 1 month.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.