Learn More
Medchemexpress LLC Dope-peg-azide | 99.6% | 1000 (average) | 5 MG
SDP
Catalog No. 5000284062
Encompass_Preferred
DOPE-PEG-Azide (MW 1000) is a DOPE-based phospholipid functionalized with a polyethylene glycol (PEG) chain terminating in an azide group. The azide handle enables bioorthogonal click chemistry for conjugation and surface modification of liposomes, lipid nanoparticles, and other lipid-based formulations. The material is polydisperse with an average PEG molecular weight of 1000 and is supplied for research use.
  • Provides azide functional group for copper-catalyzed or copper-free click chemistry.
  • Average PEG molecular weight 1000 (polydisperse).
  • High purity suitable for formulation and bioconjugation (purity listed as 99.62%).
  • Suitable for liposome and lipid nanoparticle functionalization.
  • Available in small pack sizes for screening and method development.
  • Store at -20 °C in pure form.
Catalog No. 50-002-84062 Supplier Medchemexpress LLC Supplier No. HYW5914485MG
DOPE-PEG-Azide (MW 1000) is a DOPE-based phospholipid functionalized with a polyethylene glycol (PEG) chain terminating in an azide group. The azide handle enables bioorthogonal click chemistry for conjugation and surface modification of liposomes, lipid nanoparticles, and other lipid-based formulations. The material is polydisperse with an average PEG molecular weight of 1000 and is supplied for research use.
  • Provides azide functional group for copper-catalyzed or copper-free click chemistry.
  • Average PEG molecular weight 1000 (polydisperse).
  • High purity suitable for formulation and bioconjugation (purity listed as 99.62%).
  • Suitable for liposome and lipid nanoparticle functionalization.
  • Available in small pack sizes for screening and method development.
  • Store at -20 °C in pure form.