Learn More
Medchemexpress LLC DOPE-PEG-azide (MW 3400) | 99.8% | 3400 (average) | 100 MG
SDP
Catalog No. 5000284020
Encompass_Preferred
DOPE-PEG-Azide is a PEGylated phospholipid bearing an azide functional group on the PEG terminus (average molecular weight 3,400 Da). It is supplied as a powder for research use and is intended for incorporation into liposomes, lipid nanoparticles, and surface-modification workflows where the azide terminus enables bioorthogonal conjugation.
Catalog No. 50-002-84020 Supplier Medchemexpress LLC Supplier No. HYW591450100MG
DOPE-PEG-Azide is a PEGylated phospholipid bearing an azide functional group on the PEG terminus (average molecular weight 3,400 Da). It is supplied as a powder for research use and is intended for incorporation into liposomes, lipid nanoparticles, and surface-modification workflows where the azide terminus enables bioorthogonal conjugation.