Learn More
Medchemexpress LLC Dope-PEG-COOH | 95.0% | 2000 Da | 100 MG
SDP
Catalog No. 5000281273
Encompass_Preferred
DOPE-PEG-COOH is a lipid-polyethylene glycol (PEG) conjugate in which a dioleoylphosphatidylethanolamine lipid is linked to a carboxyl-terminated PEG chain with an average molecular weight of about 2,000 Da. It is intended for research use to prepare liposomes, pegylated nanoparticles, and surface-modified formulations where a reactive carboxylic acid group is needed for further conjugation. The product is polydisperse and is typically dissolved in DMSO for handling.
  • Average molecular weight ~2000 Da.
  • Carboxyl terminal suitable for bioconjugation.
  • Enables liposome and nanoparticle formulation.
  • Amphiphilic lipid tail for hydrophobic payload incorporation.
  • PEG chain provides aqueous solubility and steric stabilization.
  • Typical purity around 95.0%.
  • Soluble in DMSO; handle with freshly opened solvent.
  • Store pure material at low temperatures per supplier guidance.
Catalog No. 50-002-81273 Supplier Medchemexpress LLC Supplier No. HYW440827100MG
DOPE-PEG-COOH is a lipid-polyethylene glycol (PEG) conjugate in which a dioleoylphosphatidylethanolamine lipid is linked to a carboxyl-terminated PEG chain with an average molecular weight of about 2,000 Da. It is intended for research use to prepare liposomes, pegylated nanoparticles, and surface-modified formulations where a reactive carboxylic acid group is needed for further conjugation. The product is polydisperse and is typically dissolved in DMSO for handling.
  • Average molecular weight ~2000 Da.
  • Carboxyl terminal suitable for bioconjugation.
  • Enables liposome and nanoparticle formulation.
  • Amphiphilic lipid tail for hydrophobic payload incorporation.
  • PEG chain provides aqueous solubility and steric stabilization.
  • Typical purity around 95.0%.
  • Soluble in DMSO; handle with freshly opened solvent.
  • Store pure material at low temperatures per supplier guidance.