Learn More
Medchemexpress LLC Dope-peg-dbco | 90.0% | 2000 | 5 MG
SDP
Catalog No. 5000391036
Encompass_Preferred
DOPE-PEG-DBCO (MW 2000) is a PEG lipid that includes DOPE and DBCO as functional groups. DOPE serves as a helper lipid in gene transfection, PEG is utilized in drug delivery systems, and DBCO can be employed for click chemistry.
  • Helper lipid (DOPE) for gene transfection
  • Used in drug delivery systems (PEG)
  • Suitable for click chemistry (DBCO)
  • Appearance: Oil
  • Color: White to off-white
Catalog No. 50-003-91036 Supplier Medchemexpress LLC Supplier No. HY1640355MG
DOPE-PEG-DBCO (MW 2000) is a PEG lipid that includes DOPE and DBCO as functional groups. DOPE serves as a helper lipid in gene transfection, PEG is utilized in drug delivery systems, and DBCO can be employed for click chemistry.
  • Helper lipid (DOPE) for gene transfection
  • Used in drug delivery systems (PEG)
  • Suitable for click chemistry (DBCO)
  • Appearance: Oil
  • Color: White to off-white