Learn More
Medchemexpress LLC Dope-peg-mal | 98.2% | 2000 Da | 5 MG
SDP
Catalog No. 5000280978
Encompass_Preferred
DOPE-PEG-Mal (MW 2000) is a maleimide-functionalized dioleoyl phosphoethanolamine-polyethylene glycol conjugate formulated for liposome and nanoparticle preparation and for covalent attachment to thiol-containing ligands. The maleimide group enables rapid, selective thiol coupling under mild conditions to yield stable thioether linkages; the PEG moiety (average MW ~2000 Da) provides steric stabilization in formulations.
  • Maleimide-functionalized lipid for thiol conjugation.
  • Selective thiol reactivity at pH 6.5-7.5 forming stable thioether bonds.
  • Suitable for liposome and nanoparticle formulation.
  • Average PEG molecular weight ~2000 Da for steric stabilization.
  • High reported purity (≈98.2%).
  • Available in small research pack sizes for formulation work.
Catalog No. 50-002-80978 Supplier Medchemexpress LLC Supplier No. HYW4409955MG
DOPE-PEG-Mal (MW 2000) is a maleimide-functionalized dioleoyl phosphoethanolamine-polyethylene glycol conjugate formulated for liposome and nanoparticle preparation and for covalent attachment to thiol-containing ligands. The maleimide group enables rapid, selective thiol coupling under mild conditions to yield stable thioether linkages; the PEG moiety (average MW ~2000 Da) provides steric stabilization in formulations.
  • Maleimide-functionalized lipid for thiol conjugation.
  • Selective thiol reactivity at pH 6.5-7.5 forming stable thioether bonds.
  • Suitable for liposome and nanoparticle formulation.
  • Average PEG molecular weight ~2000 Da for steric stabilization.
  • High reported purity (≈98.2%).
  • Available in small research pack sizes for formulation work.