Learn More
Medchemexpress LLC Dope-PEG-Mal | 98.2% | 2000 Da (average) | 10 MG
SDP
Catalog No. 5000281009
Encompass_Preferred
DOPE-PEG-Mal (MW 2000) is a maleimide-functionalized 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine conjugated to polyethylene glycol with an average molecular weight of 2,000 Da. The maleimide group selectively reacts with thiol groups at pH 6.5-7.5 to form stable thioether bonds, enabling covalent attachment of thiol-containing ligands. Supplied as a solid for use in liposome and nanoparticle preparation and for surface functionalization in bioconjugation research.
  • Maleimide-functionalized PEGylated phospholipid.
  • Average molecular weight 2,000 Da.
  • Reacts with thiols at pH 6.5-7.5 to form stable thioether bonds.
  • Suitable for liposome and nanoparticle formulation.
  • Supplied as a solid in small research quantities.
  • Certificate of analysis and safety data sheet available.
Catalog No. 50-002-81009 Supplier Medchemexpress LLC Supplier No. HYW44099510MG
DOPE-PEG-Mal (MW 2000) is a maleimide-functionalized 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine conjugated to polyethylene glycol with an average molecular weight of 2,000 Da. The maleimide group selectively reacts with thiol groups at pH 6.5-7.5 to form stable thioether bonds, enabling covalent attachment of thiol-containing ligands. Supplied as a solid for use in liposome and nanoparticle preparation and for surface functionalization in bioconjugation research.
  • Maleimide-functionalized PEGylated phospholipid.
  • Average molecular weight 2,000 Da.
  • Reacts with thiols at pH 6.5-7.5 to form stable thioether bonds.
  • Suitable for liposome and nanoparticle formulation.
  • Supplied as a solid in small research quantities.
  • Certificate of analysis and safety data sheet available.