Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dope-peg-mal | CAS | MFCD | 98.2% | 2000 (average) | 50 MG
SDP

Supplier:  Medchemexpress LLC HYW44099550MG

Encompass_Preferred

DOPE-PEG-Mal (MW 2000) is a phospholipid-polyethylene glycol conjugate designed for preparing liposomes and nanoparticles and for thiol-specific bioconjugation. The maleimide functional group reacts with thiols at pH 6.5-7.5 to form stable thioether bonds. Supplied as a solid with high purity for research applications.

  • Phospholipid-PEG conjugate suitable for liposome and nanoparticle formulation.
  • Maleimide group reacts with thiols at pH 6.5-7.5.
  • Average molecular weight 2000.
  • Purity 98.2%.
  • Soluble in DMSO (~100 mg/mL; may require ultrasonic treatment).
  • Powder stable at -20 °C for extended storage.
  • Intended for research use only.

Catalog No. 50-002-81710


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.