Learn More
Kyfora Bio DOPE 250 MG
Catalog No. 502559772
Encompass_Preferred
DOPE (1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine) is a neutral "helper" phospholipid commonly utilized in liposomal and lipid nanoparticle (LNP) formulations for gene and drug delivery. When paired with cationic or ionizable lipids, DOPE enhances cellular uptake and fusion efficiency, improving the delivery of liposomes and LNPs across a range of cell types. This helper lipid is essential for optimizing formulation performance in both in vitro and in vivo applications. For research use only.
Catalog No. 50-255-9772 Supplier Kyfora Bio Supplier No. 26451250
DOPE (1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine) is a neutral "helper" phospholipid commonly utilized in liposomal and lipid nanoparticle (LNP) formulations for gene and drug delivery. When paired with cationic or ionizable lipids, DOPE enhances cellular uptake and fusion efficiency, improving the delivery of liposomes and LNPs across a range of cell types. This helper lipid is essential for optimizing formulation performance in both in vitro and in vivo applications. For research use only.