Learn More
Medchemexpress LLC Dopexamine hydrochloride | 86484-91-5 | 429.42 | 25 MG
SDP
Catalog No. 5000448100
Encompass_Preferred
Dopexamine hydrochloride is a β2 adrenergic receptor agonist and a dopamine analogue. It exhibits agonist activity at β2 and dopaminergic receptors, attenuating systemic inflammatory response, reducing tissue leukocyte infiltration, and protecting against organ injury. It can reduce the systemic inflammatory response to endotoxin, including cytokine release, endothelial adhesion molecules, and oxidative stress, without significantly altering systemic hemodynamics.
  • Agonist activity at β2 and dopaminergic receptors
  • Attenuates systemic inflammatory response
  • Reduces tissue leukocyte infiltration
  • Protects against organ injury
  • Stimulates β-adrenergic and dopamine 1 and 2 receptors
  • Confers vasodilatory effects
Catalog No. 50-004-48100 Supplier Medchemexpress LLC Supplier No. 25MGHYU00205
Dopexamine hydrochloride is a β2 adrenergic receptor agonist and a dopamine analogue. It exhibits agonist activity at β2 and dopaminergic receptors, attenuating systemic inflammatory response, reducing tissue leukocyte infiltration, and protecting against organ injury. It can reduce the systemic inflammatory response to endotoxin, including cytokine release, endothelial adhesion molecules, and oxidative stress, without significantly altering systemic hemodynamics.
  • Agonist activity at β2 and dopaminergic receptors
  • Attenuates systemic inflammatory response
  • Reduces tissue leukocyte infiltration
  • Protects against organ injury
  • Stimulates β-adrenergic and dopamine 1 and 2 receptors
  • Confers vasodilatory effects