Learn More
Medchemexpress LLC HY-10320 100mg Medchemexpress, Doramapimod CAS:285983-48-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10320100MG
Encompass_Preferred
Medchemexpress, HY-10320 100mg Doramapimod CAS:285983-48-4 Doramapimod (BIRB 796) is an orally active, highly potent p38 MAPK inhibitor, which has an IC50 for p38α=38 nM, for p38β=65 nM, for p38γ=200 nM, and for p38δ=520 nM. Doramapimod has picomolar affinity for p38 kinase (Kd=0.1 nM). Doramapimod also inhibits B-Raf with an IC50 of 83 nM[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8724