Learn More
Selleck Chemical LLC Doramectin S5117-1g
SDP
Catalog No. 503110186
Encompass_Preferred
Doramectin (Cyclohexylavermectin B1 Dectomax) is a macrocyclic lactone derivative of ivermectin an anthelmintic that potentiates glutamate- and GABA-gated chloride channel opening in nematodes It is used for the treatment and control of internal parasitosis ticks and mange
Catalog No. 50-311-0186 Supplier Selleck Chemical LLC Supplier No. S51171G
Doramectin (Cyclohexylavermectin B1 Dectomax) is a macrocyclic lactone derivative of ivermectin an anthelmintic that potentiates glutamate- and GABA-gated chloride channel opening in nematodes It is used for the treatment and control of internal parasitosis ticks and mange