Learn More
Selleck Chemical LLC Doravirine
SDP
Catalog No. 5000003316
Encompass_Preferred
Doravirine is a novel HIV-1 nonnucleoside reverse transcriptase inhibitor with IC50 values of 12 9 7 and 9 7 nM against the wild type (WT) and K103N and Y181C reverse transcriptase (RT) mutants respectively in a biochemical assay It is highly specific with minimum off-target activities
Catalog No. 50-000-03316 Supplier Selleck Chemical LLC Supplier No. S64922MG
Doravirine is a novel HIV-1 nonnucleoside reverse transcriptase inhibitor with IC50 values of 12 9 7 and 9 7 nM against the wild type (WT) and K103N and Y181C reverse transcriptase (RT) mutants respectively in a biochemical assay It is highly specific with minimum off-target activities