Learn More
ATCC Agrobacterium fabrum; Strain NTL4(pZLR4)
Catalog No. 502381621
Encompass
Agrobacterium fabrum strain NTL4(pZLR4) was derived from strain NTL4 via the transformation of recombinant plasmid pZLR4. This strain can be used as a bioreporter for detecting N-acyl homoserine lactone quorum-sensing signals.
Catalog No. 50-238-1621 Supplier ATCC Supplier No. BAA2240
Add to Cart
Add to Cart
Agrobacterium fabrum strain NTL4(pZLR4) was derived from strain NTL4 via the transformation of recombinant plasmid pZLR4. This strain can be used as a bioreporter for detecting N-acyl homoserine lactone quorum-sensing signals.