Learn More
Alkali Scientific Doripenem, 1 G
SDP
Catalog No. 5000660337
Encompass_Preferred
Doripenem is a broad-spectrum antibiotic used in clinical and laboratory research to treat infections caused by Gram-positive and Gram-negative bacteria. The 1g quantity is commonly used in antimicrobial susceptibility testing, bacterial resistance studies, and the development of new antibiotics. Doripenem inhibits bacterial cell wall synthesis and is effective against a wide variety of pathogens. It is especially valuable in studies investigating carbapenem-resistant organisms. This product plays a critical role in exploring bacterial pathogenicity, developing new therapeutic approaches, and understanding the global challenge of antibiotic resistance.
Catalog No. 50-006-60337 Supplier Alkali Scientific Supplier No. DOR003CP
Doripenem is a broad-spectrum antibiotic used in clinical and laboratory research to treat infections caused by Gram-positive and Gram-negative bacteria. The 1g quantity is commonly used in antimicrobial susceptibility testing, bacterial resistance studies, and the development of new antibiotics. Doripenem inhibits bacterial cell wall synthesis and is effective against a wide variety of pathogens. It is especially valuable in studies investigating carbapenem-resistant organisms. This product plays a critical role in exploring bacterial pathogenicity, developing new therapeutic approaches, and understanding the global challenge of antibiotic resistance.