Learn More
TARGETMOL CHEMICALS INC DORIPENEM 50MG
SDP
Catalog No. 502577192
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Doripenem is a new parenteral carbapenem antibiotic with broad-spectrum activity against gram-negative and gram-positive pathogens. purity: 99%
Catalog No. 50-257-7192 Supplier TARGETMOL CHEMICALS INC Supplier No. T2231450MG
Also available in 5 mg 10 mg 25 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Doripenem is a new parenteral carbapenem antibiotic with broad-spectrum activity against gram-negative and gram-positive pathogens. purity: 99%