Learn More
Selleck Chemical LLC Doripenem Hydrate 50mg 364622-82-2 S-4661
SDP
Catalog No. 506766
Encompass_Preferred
Doripenem Hydrate (S-4661) is an ultra-broad-spectrum injectable antibiotic, used to treat complicated intra-abdominal infections and complicated urinary tract infections. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-676-6 Supplier Selleck Chemical LLC Supplier No. S137450MG
Doripenem Hydrate (S-4661) is an ultra-broad-spectrum injectable antibiotic, used to treat complicated intra-abdominal infections and complicated urinary tract infections. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.