Learn More
Cayman Chemical Doripenem-d4 (sodium salt), Purity: ≥99% deuterated forms (d1-d4), 1 mg
SDP
Catalog No. 502934574
Encompass_Preferred
An internal standard for the quantification of doripenem by GC- or LC-MS
Catalog No. 50-293-4574 Supplier Cayman Chemical Supplier No. 265221 MG
An internal standard for the quantification of doripenem by GC- or LC-MS