Learn More
Cayman Chemical DorIpenm-d4sodIum salt 500ug
SDP
Supplier:  Cayman Chemical 26522500 UG
Encompass_Preferred
An internal standard for the quantification of doripenem by GC- or LC-MS
Catalog No. 50-293-4380