Learn More
Cayman Chemical Doripenem (hydrate), Purity: ≥98%, CAS: 1820954-21-9, 10 mg
SDP
Catalog No. 502979873
Encompass_Preferred
A broad-spectrum antibiotic; is active against Gram-negative and Gram-positive bacteria, including S. aureus, S. pneumoniae, E. coli, and K. pneumoniae (MICs = 0.03-0.06, 0.016-0.06, P. aeruginosa respiratory tract infection at 100 mg/kg per day
Catalog No. 50-297-9873 Supplier Cayman Chemical Supplier No. 1693410 MG
A broad-spectrum antibiotic; is active against Gram-negative and Gram-positive bacteria, including S. aureus, S. pneumoniae, E. coli, and K. pneumoniae (MICs = 0.03-0.06, 0.016-0.06, P. aeruginosa respiratory tract infection at 100 mg/kg per day