Learn More
MSC Dormer Pramet Straight Flute Tap, 1-1/16-12 UN, HSS, 1 ea
Catalog No. NC3845982
Encompass
This machine straight flute tap is designed for precise internal threading applications. Constructed from high-speed steel, it is suitable for creating 1-1/16-12 UN threads with a 2B class of fit in various materials.
  • Thread size 1-1/16-12 UN with 2B class of fit
  • Four flutes for efficient chip evacuation
  • Bottoming chamfer for threading blind holes
  • High-speed steel construction for durability and wear resistance
  • Bright/uncoated finish for general purpose tapping
  • Right-hand thread direction
  • Suitable for applications in blind and through-holes
  • Compatible with a wide range of materials including steel, cast iron, aluminum, and plastics.
  • Overall length of 127.00 mm and thread length of 37.00 mm
Catalog No. NC3845982 Supplier MSC Supplier No. 65685281
This machine straight flute tap is designed for precise internal threading applications. Constructed from high-speed steel, it is suitable for creating 1-1/16-12 UN threads with a 2B class of fit in various materials.
  • Thread size 1-1/16-12 UN with 2B class of fit
  • Four flutes for efficient chip evacuation
  • Bottoming chamfer for threading blind holes
  • High-speed steel construction for durability and wear resistance
  • Bright/uncoated finish for general purpose tapping
  • Right-hand thread direction
  • Suitable for applications in blind and through-holes
  • Compatible with a wide range of materials including steel, cast iron, aluminum, and plastics.
  • Overall length of 127.00 mm and thread length of 37.00 mm