Learn More
Med Vet International Dorsi-Flex Assist, Medium, J1005B, Each
Supplier: Med Vet International JOR1005B
Provides light to moderate assistance for normal hind paw placement. Designed for use as a therapy aid or for daily activity. (Not to be used for dogs with severe neurological impairments.) Reduces knuckling/standing on the dorsum of the paw Mobility aid for unstable or weak distal hind legs Assists with tarsal/hock flexion and digital extension Can be used bilaterally Features Adjustable, elastic tension straps Includes breathable, anti-skid, Thera-Paw boot Will not immobilize joints Lightweight Easy to put on/take off Cleans easily by rinsing with warm water Medium Includes Thera-Paw size M/L #2 (paw 3"W x 3 3/4in.L) and Tibial cuff up to 6 1/2in. circumference. NOTE: Due to risk of infection, all splints should be applied only on limbs of animals with protective covering. We will not accept any splint returned containing animal hair.