Learn More
BIOGEMS INTERNATIONAL INC Dorsomorphin 10mg
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 866643010MG
Encompass
Dorsomorphin is a potent and selective inhibitor of the BMP pathway through the ALK2, ALK3, and ALK6 receptors. It is also a selective and reversible inhibitor of the AMP-activated protein kinase (AMPK). Dorsomorphin is reported to be useful in promoting specific cell differentiation and inducing cancer cell line autophagy.
Catalog No. 50-153-0511