Learn More
Medchemexpress LLC HY-13418A 10mg Medchemexpress, Dorsomorphin CAS:866405-64-3 Purity:>98%
SDP
Catalog No. 502028991
Encompass_Preferred
Medchemexpress, HY-13418A 10mg Dorsomorphin CAS:866405-64-3 Dorsomorphin (Compound C) is a selective and ATP-competitive AMPK inhibitor (Ki=109 nM in the absence of AMP). Dorsomorphin (BML-275) selectively inhibits BMP type I receptors ALK2, ALK3, and ALK6. Dorsomorphin induces autophagy[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8991 Supplier Medchemexpress LLC Supplier No. HY13418A10MG
Add to Cart
Add to Cart
Medchemexpress, HY-13418A 10mg Dorsomorphin CAS:866405-64-3 Dorsomorphin (Compound C) is a selective and ATP-competitive AMPK inhibitor (Ki=109 nM in the absence of AMP). Dorsomorphin (BML-275) selectively inhibits BMP type I receptors ALK2, ALK3, and ALK6. Dorsomorphin induces autophagy[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.