Learn More
Selleck Chemical LLC Dorsomorphin (Compound C) 2HCl 10mg 1219168-18-9 BML-275 2HCl,Compound C 2HCl
SDP
Supplier:  Selleck Chemical LLC S730610MG
Encompass_Preferred
Dorsomorphin 2HCl (BML-275, Compound C) is a potent, reversible, selective AMPK inhibitor with Ki of 109 nM in cell-free assays, exhibiting no significant inhibition of several structurally related kinases including ZAPK, SYK, PKC?, PKA, and JAK3. Also inhibits type ? BMP receptor activity. Dorsomorphin induces autophagy in cancer cell line. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1058056