Learn More
Apexbio Technology LLC Dorsomorphin 2HCl, 10mg. Cas: 1219168-18-9 MFCD: MFCD11112197. AMPK inhibitor
SDP
Supplier:  Apexbio Technology LLC B137210
Encompass_Preferred
Dorsomorphin is a potent, reversible, selective AMPK inhibitor with K i of 109 nM, exhibiting no significant inhibition of several structurally related kinases including ZAPK, SYK, PKC, PKA, and JAK3. 1219168-18-9. MFCD11112197
Catalog No. 50-101-3624