Learn More
Selleck Chemical LLC Dorsomorphin 2HCl
SDP
Supplier:  Selleck Chemical LLC S7306200MG
Encompass_Preferred
Dorsomorphin 2HCl is a potent reversible selective AMPK inhibitor with Ki of 109 nM in cell-free assays exhibiting no significant inhibition of several structurally related kinases including ZAPK SYK PKC PKA and JAK3 Also inhibits type BMP receptor activity Dorsomorphin induces autophagy in cancer cell line
Catalog No. 50-000-04476