Learn More
Selleck Chemical LLC Dorsomorphin 2HCl
SDP
Catalog No. 5000004635
Encompass_Preferred
Dorsomorphin 2HCl is a potent reversible selective AMPK inhibitor with Ki of 109 nM in cell-free assays exhibiting no significant inhibition of several structurally related kinases including ZAPK SYK PKC PKA and JAK3 Also inhibits type BMP receptor activity Dorsomorphin induces autophagy in cancer cell line
Catalog No. 50-000-04635 Supplier Selleck Chemical LLC Supplier No. S73061G
Add to Cart
Add to Cart
Dorsomorphin 2HCl is a potent reversible selective AMPK inhibitor with Ki of 109 nM in cell-free assays exhibiting no significant inhibition of several structurally related kinases including ZAPK SYK PKC PKA and JAK3 Also inhibits type BMP receptor activity Dorsomorphin induces autophagy in cancer cell line