Learn More
Apexbio Technology LLC Dorsomorphin 2HCl 1219168-18-9 50mg
SDP
Catalog No. 5000062066
Encompass_Preferred
Dorsomorphin 2HCl (CAS 1219168-18-9) is a small-molecule inhibitor targeting BMP signaling through selective blockade of BMP type I receptors ALK2 ALK3 and ALK6 It prevents phosphorylation of downstream effectors SMAD1/5/8 with an IC50 of approximately 0 47 mM Originally identified by zebrafish phenotypic screening as a disruptor of embryonic dorsoventral patterning Dorsomorphin has been employed to dissect BMP-regulated pathways in iron homeostasis significantly affecting hepcidin expression and iron metabolism in vitro and in vivo It is widely utilized in mechanistic studies of BMP function and signaling regulation
Catalog No. 50-000-62066 Supplier Apexbio Technology LLC Supplier No. B137250
Dorsomorphin 2HCl (CAS 1219168-18-9) is a small-molecule inhibitor targeting BMP signaling through selective blockade of BMP type I receptors ALK2 ALK3 and ALK6 It prevents phosphorylation of downstream effectors SMAD1/5/8 with an IC50 of approximately 0 47 mM Originally identified by zebrafish phenotypic screening as a disruptor of embryonic dorsoventral patterning Dorsomorphin has been employed to dissect BMP-regulated pathways in iron homeostasis significantly affecting hepcidin expression and iron metabolism in vitro and in vivo It is widely utilized in mechanistic studies of BMP function and signaling regulation