Learn More
Selleck Chemical LLC Dorsomorphin
SDP
Catalog No. 5000001548
Encompass_Preferred
Dorsomorphin is a potent reversible selective AMPK inhibitor with Ki of 109 nM in cell-free assays exhibiting no significant inhibition of several structurally related kinases including ZAPK SYK PKC PKA and JAK3 Dorsomorphin selectively inhibits the BMP type I receptors ALK2 ALK3 and ALK6 Dorsomorphin is used in promoting specific cell differentiation and inducing cancer cell line autophagy For cell testing the water-soluble S7306 Dorsomorphin (Compound C) 2HCl is recommended
Catalog No. 50-000-01548 Supplier Selleck Chemical LLC Supplier No. S7840100MG
Dorsomorphin is a potent reversible selective AMPK inhibitor with Ki of 109 nM in cell-free assays exhibiting no significant inhibition of several structurally related kinases including ZAPK SYK PKC PKA and JAK3 Dorsomorphin selectively inhibits the BMP type I receptors ALK2 ALK3 and ALK6 Dorsomorphin is used in promoting specific cell differentiation and inducing cancer cell line autophagy For cell testing the water-soluble S7306 Dorsomorphin (Compound C) 2HCl is recommended