Learn More
Cayman Chemical Dorsomorphin, Purity: ≥98%, CAS: 866405-64-3, 50 mg
SDP
Catalog No. 502952558
Encompass_Preferred
A potent, reversible inhibitor of AMPK (Ki = 109 nM); dose-dependently inhibits the bone morphogenetic protein type 1 receptors ACTR-I (ALK2), BMPR-IA (ALK3), and BMPR-IB (ALK6); downregulates the Akt/mTOR pathway to induce autophagy in U251 human glioma cells
Catalog No. 50-295-2558 Supplier Cayman Chemical Supplier No. 1196750 MG
A potent, reversible inhibitor of AMPK (Ki = 109 nM); dose-dependently inhibits the bone morphogenetic protein type 1 receptors ACTR-I (ALK2), BMPR-IA (ALK3), and BMPR-IB (ALK6); downregulates the Akt/mTOR pathway to induce autophagy in U251 human glioma cells