Learn More
Apexbio Technology LLC Dorsomorphin (Compound C), 866405-64-3, MFCD08705402, 5 mg
SDP
Catalog No. 501151641
Encompass_Preferred
MF: C24H25N5O, MW: 399.49, Solubility: 8.49 mg/mL in DMSO with ultrasonic and warming,insoluble in H2O, Synonyms: Compound C. Dorsomorphin is a cell-permeable and reversible ATP-competitive inhibitor of AMP-activated protein kinase (AMPK) with Ki value of 109nM.
Catalog No. 50-115-1641 Supplier Apexbio Technology LLC Supplier No. B32525
MF: C24H25N5O, MW: 399.49, Solubility: 8.49 mg/mL in DMSO with ultrasonic and warming,insoluble in H2O, Synonyms: Compound C. Dorsomorphin is a cell-permeable and reversible ATP-competitive inhibitor of AMP-activated protein kinase (AMPK) with Ki value of 109nM.