Learn More
Abcam Dorsomorphin (Compound C), AMP-kinase inhibitor, 10MG
Catalog No. 502584561
Encompass_Preferred
MW 399.5 Da, Purity >99%. Potent, selective and reversible inhibitor of AMP-kinase (Ki = 109 nM). Inhibits AMPK activation induced by AICAR (ab120358) and metformin (ab120847). Selective inhibitor of bone morphogenetic protein (BMP) type I receptors (ALK2, ALK3 and ALK6). Does not affect ZAPK, SYK, PKCθ, PKA and JAK3. Cell-permeable. Also available in a water soluble form (Dorsomorphin dihydrochloride - ab144821).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4561 Supplier Abcam Supplier No. AB12084310MG
MW 399.5 Da, Purity >99%. Potent, selective and reversible inhibitor of AMP-kinase (Ki = 109 nM). Inhibits AMPK activation induced by AICAR (ab120358) and metformin (ab120847). Selective inhibitor of bone morphogenetic protein (BMP) type I receptors (ALK2, ALK3 and ALK6). Does not affect ZAPK, SYK, PKCθ, PKA and JAK3. Cell-permeable. Also available in a water soluble form (Dorsomorphin dihydrochloride - ab144821).
The product is subject to the following: Abcam Restricted Use Statement