Learn More
Medchemexpress LLC HY-13418 10mg Medchemexpress, Dorsomorphin (dihydrochloride) CAS:1219168-18-9 Purity:>98%
SDP
Catalog No. 502028990
Encompass_Preferred
Medchemexpress, HY-13418 10mg Dorsomorphin (dihydrochloride) CAS:1219168-18-9 Dorsomorphin dihydrochloride (BML-275 dihydrochloride; Compound C dihydrochloride) is a potent, selective and ATP-competitive AMPK inhibitor, with a Ki of 109 nM. Dorsomorphin dihydrochloride inhibits BMP pathway by targeting the type I receptors ALK2, ALK3, and ALK6. Dorsomorphin dihydrochloride induces autophagy[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8990 Supplier Medchemexpress LLC Supplier No. HY1341810MG
Medchemexpress, HY-13418 10mg Dorsomorphin (dihydrochloride) CAS:1219168-18-9 Dorsomorphin dihydrochloride (BML-275 dihydrochloride; Compound C dihydrochloride) is a potent, selective and ATP-competitive AMPK inhibitor, with a Ki of 109 nM. Dorsomorphin dihydrochloride inhibits BMP pathway by targeting the type I receptors ALK2, ALK3, and ALK6. Dorsomorphin dihydrochloride induces autophagy[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.