Learn More
Enzo Life Sciences Dorsomorphin hydrochloride (5mg). CAS: 1219168-18-9
SDP
Catalog No. 502013441
Encompass_Preferred
Potent and selective AMPK inhibitor. Dorsomorphin also inhibits the bone morphogenetic protein type 1 receptors ACTR-I (ALK-2), BMPR-IA (ALK-3), and BMPR-IB (ALK-6). Alternative name: 6-[4-[2-(1-Piperidinyl)ethoxy]phenyl]-3-(4-pyridinyl)-pyrazolo[1,5-a]pyrimidine dihydrochloride. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO or water. Long Term Storage: -80°C.
Catalog No. 50-201-3441 Supplier Enzo Life Sciences Supplier No. ENZCHM1410005
Potent and selective AMPK inhibitor. Dorsomorphin also inhibits the bone morphogenetic protein type 1 receptors ACTR-I (ALK-2), BMPR-IA (ALK-3), and BMPR-IB (ALK-6). Alternative name: 6-[4-[2-(1-Piperidinyl)ethoxy]phenyl]-3-(4-pyridinyl)-pyrazolo[1,5-a]pyrimidine dihydrochloride. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO or water. Long Term Storage: -80°C.