Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dorsomorphin 97.0+%, TCI America™
SDP

Catalog No. D539410MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D539410MG 10 mg
D539450MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D539410MG Supplier TCI America Supplier No. D539410MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 866405-64-3
Molecular Formula C24H25N5O
Molecular Weight (g/mol) 399.50
MDL Number MFCD08705402
InChI Key XHBVYDAKJHETMP-UHFFFAOYSA-N
Synonym 6-[4-(2-Piperidin-1-ylethoxy)phenyl]-3-pyridin-4-ylpyrazolo[1,5-a]pyrimidine
PubChem CID 11524144
ChEBI CHEBI:78510
IUPAC Name 4-(6-{4-[2-(piperidin-1-yl)ethoxy]phenyl}pyrazolo[1,5-a]pyrimidin-3-yl)pyridine
SMILES C(CN1CCCCC1)OC1=CC=C(C=C1)C1=CN2N=CC(=C2N=C1)C1=CC=NC=C1

Specifications

Quantity 10 mg
Formula Weight 399.50
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Dorsomorphin
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.